missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, ACS Reagent, Reagents

Catalog No. 01337710
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337710 2.5 kg
01337712 100 g
01337711 10 kg
01337709 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337710 Supplier Labchem Inc Supplier No. C13205002.5A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Nitrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym Nitrate of Soda
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380°C
Melting Point 306°C
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Percent Purity ≥99%
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.26
Chemical Name or Material Sodium Nitrate, ACS Reagent