missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-Octanoate 99.0+%, TCI America™

Catalog No. p-7134143
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003425G 25 g
O0034250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003425G Supplier TCI America Supplier No. O003425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1984-06-1
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD00058511
InChI Key BYKRNSHANADUFY-UHFFFAOYSA-M
Synonym sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, caprylic acid sodium salt, natrium octanoat, octanoic acid sodium salt, unii-9xtm81vk2b, 9xtm81vk2b
PubChem CID 23664772
IUPAC Name sodium;octanoate
SMILES CCCCCCCC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 166.20
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium n-Octanoate
EINECSNumber (2)-0611&(7)-0973
RTECSNumber RH0787000
TSCA Yes