missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium N-Lauroylsarcosinate 98.0+%, TCI America™
$32.28 - $134.80
Chemical Identifiers
| CAS | 137-16-6 |
|---|---|
| Molecular Formula | C15H28NNaO3 |
| Molecular Weight (g/mol) | 293.38 |
| MDL Number | MFCD00042728 |
| InChI Key | KSAVQLQVUXSOCR-UHFFFAOYSA-M |
| Synonym | N-Dodecanoylsarcosine Sodium Salt, N-Lauroylsarcosine Sodium Salt, Sodium N-Dodecanoylsarcosinate |
| PubChem CID | 23668817 |
| IUPAC Name | sodium 2-(N-methyldodecanamido)acetate |
| SMILES | [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O |
Chemical Identifiers
| C15H28NNaO3 | |
| MFCD00042728 | |
| N-Dodecanoylsarcosine Sodium Salt, N-Lauroylsarcosine Sodium Salt, Sodium N-Dodecanoylsarcosinate | |
| sodium 2-(N-methyldodecanamido)acetate |
Specifications
| White | |
| Crystal-Powder at 20°C | |
| 8.0 to 9.5 (50 g/L H2O 25°C) | |
| MFCD00042728 | |
| N-Dodecanoylsarcosine Sodium Salt, N-Lauroylsarcosine Sodium Salt, Sodium N-Dodecanoylsarcosinate | |
| [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O | |
| 293.38 | |
| 293.38 | |
| Sodium N-Lauroylsarcosinate |
| 137-16-6 | |
| 46°C | |
| C15H28NNaO3 | |
| 2811 | |
| KSAVQLQVUXSOCR-UHFFFAOYSA-M | |
| sodium 2-(N-methyldodecanamido)acetate | |
| 23668817 | |
| ≥98.0% (N) |
Safety and Handling
EINECSNumber : (2)-1226
RTECSNumber : MC0598960
TSCA : Yes