missing translation for 'onlineSavingsMsg'
Learn More

Sodium N-Lauroylsarcosinate 98.0+%, TCI America™

Catalog No. L0019500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0019500G 500 g
L001925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0019500G Supplier TCI America Supplier No. L0019500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym N-Dodecanoylsarcosine Sodium Salt, N-Lauroylsarcosine Sodium Salt, Sodium N-Dodecanoylsarcosinate
PubChem CID 23668817
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Melting Point 46°C
pH 8.0 to 9.5 (50 g/L H2O 25°C)
Quantity 500 g
UN Number 2811
Formula Weight 293.38
Percent Purity ≥98.0% (N)
Chemical Name or Material Sodium N-Lauroylsarcosinate
EINECSNumber (2)-1226
RTECSNumber MC0598960
TSCA Yes