missing translation for 'onlineSavingsMsg'
Learn More

Sodium N-Lauroylsarcosinate Hydrate 98.0+%, TCI America™

Supplier:  TCI America S059725G

 View more versions of this product

Catalog No. S059725G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium N-Lauroylsarcosinate Hydrate [for Biochemical Research]
46°C
8.0 to 9.5 (50g/L H2O 25°C)
MFCD00042728
2811
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sodium 2-(N-methyldodecanamido)acetate
23668817
≥98.0% (N)
137-16-6
White
C15H28NNaO3
25 g
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
293.38
293.38
Crystalline Powder

Chemical Identifiers

137-16-6
293.38
KSAVQLQVUXSOCR-UHFFFAOYSA-M
23668817
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
C15H28NNaO3
MFCD00042728
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
sodium 2-(N-methyldodecanamido)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1226

RTECSNumber : MC0598960

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions