missing translation for 'onlineSavingsMsg'
Learn More

Sodium N-Lauroylsarcosinate Hydrate 98.0+%, TCI America™

Catalog No. p-7135936
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05975G 5 g
S059725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05975G Supplier TCI America Supplier No. S05975G
Only null left

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Melting Point 46°C
Color White
pH 8.0 to 9.5 (50g/L H2O 25°C)
Quantity 5 g
UN Number 2811
Formula Weight 293.38
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium N-Lauroylsarcosinate Hydrate [for Biochemical Research]
EINECSNumber (2)-1226
RTECSNumber MC0598960
TSCA Yes