missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium n-Dodecyl Sulfate, Molecular biology grade, Calbiochem™,

$613.00

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym SDS, Sodium Lauryl Sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
428023500GM
View Documents Promotion Details
MilliporeSigma™
428023500GM
500 g Glass Bottle
Each for $613.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate
C12H25NaO4S
MFCD00036175
SDS, Sodium Lauryl Sulfate
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

White
C12H25NaO4S
SDS, Sodium Lauryl Sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
Molecular Biology
151-21-3
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
≥99%
Videos
SDS
Documents

Documents