missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade

Catalog No. p-7057934
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6760622 100 g
AAJ6760630 250 g
AAJ6760636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6760622 Supplier Thermo Scientific Chemicals Supplier No. J6760622
Only null left

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point ∼206°C
Color White
Quantity 100 g
UN Number UN2926
Beilstein 3599286
Merck Index 14,8636
Solubility Information Soluble in water
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Percent Purity 99%
Grade Molecular Biology
Physical Form Solid
Chemical Name or Material Sodium n-dodecyl sulfate
Show More Show Less
Hazard Category H228-H302+H332-H315-H318-H335-H500
Hazard Statement GHS H Statement
H228-H311-H302-H315-H319-H335
Flammable solid.
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 205-788-1
RTECSNumber WT1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only