missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-dodecyl sulfate, 99% (dry wt.), water <2%

Catalog No. p-7081280
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1118314 25 g
AAA1118322 100 g
AAA1118336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1118314 Supplier Thermo Scientific Chemicals Supplier No. A1118314
Only null left

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

Solubility
Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium;dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point ∼206°C
Assay Percent Range 99% (dry wt.)
Quantity 25 g
UN Number UN2926
Beilstein 3599286
Merck Index 148636
Solubility Information Slightly soluble in ethanol.
Formula Weight 288.38
Percent Purity 99%
Assay (dry wt.)
Chemical Name or Material Sodium n-dodecyl sulfate, Water <1.5%
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 1), Flammable solids (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2), Hazardous to the aquatic environment (chronic category 3), Acute toxicity (category 4)
Hazard Statement Causes serious eye damage, Flammable solid, May cause respiratory irritation, May form combustible dust concentrations in air, Causes skin irritation, Harmful to aquatic life with long lasting effects, Harmful if swallowed or if inhaled
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 205-788-1
RTECSNumber WT1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only