missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-dodecyl sulfate, 97%, for electrophoresis

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

Supplier:  Thermo Scientific Chemicals J6424122

Quantity 100 g

 View more versions of this product

Catalog No. AAJ6424122


Only null left

Description

Description

Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
C12H25NaO4S
MFCD00036175
n-Dodecyl sulfate sodium salt; Sodium lauryl sulfate
CHEBI:8984

Specifications

Sodium n-dodecyl sulfate
151-21-3
∼206°C
Ultrapure
CH3(CH2)11OSO3Na
UN2926
14,8636
Soluble in water; Slightly soluble in ethanol.
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
97%
White
Powder
100 g
C12H25NaO4S
MFCD00036175
3599286
n-Dodecyl sulfate sodium salt; Sodium lauryl sulfate
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
288.38
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H311-H302-H315-H319-H335
Flammable solid.
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302+H332-H315-H318-H335-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 205-788-1

RTECSNumber : WT1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions