missing translation for 'onlineSavingsMsg'
Learn More

Sodium N-Cyclohexylsulfamate 98.0+%, TCI America™

Catalog No. C048725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C048725G 25 g
C0487500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C048725G Supplier TCI America Supplier No. C048725G

Chemical Identifiers

CAS 139-05-9
Molecular Formula C6H12NNaO3S
Molecular Weight (g/mol) 201.22
MDL Number MFCD00003827
InChI Key UDIPTWFVPPPURJ-UHFFFAOYSA-M
Synonym sodium cyclamate, sodium cyclohexylsulfamate, sodium n-cyclohexylsulfamate, cyclamate sodium, cyclamic acid sodium salt, assugrin, ibiosuc, suessette, suestamin, sugarin
PubChem CID 23665706
ChEBI CHEBI:82431
IUPAC Name sodium N-cyclohexylsulfamate
SMILES [Na+].[O-]S(=O)(=O)NC1CCCCC1

Specifications

Color White
Quantity 25 g
Formula Weight 201.22
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium N-Cyclohexylsulfamate
EINECSNumber (3)-2480
RTECSNumber GV7350000
TSCA Yes