accessibility menu, dialog, popup

UserName

CAS RN 139-05-9

IUPAC Name: sodium N-cyclohexylsulfamate
Synonyms: sodium cyclamate molasses 环拉酸钠
Molecular Weight (g/mol): 201.22
Molecular Formula: C6H12NNaO3S
InChi Key: UDIPTWFVPPPURJ-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)NC1CCCCC1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)