missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methoxide 96.0+%, TCI America™

Catalog No. p-7135907
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0485100G 100 g
S0485500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0485100G Supplier TCI America Supplier No. S0485100G
Only null left

Chemical Identifiers

CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
UN Number 1431
Formula Weight 54.02
Percent Purity ≥96.0% (T)
Chemical Name or Material Sodium Methoxide
EINECSNumber (2)-0203
RTECSNumber PC3570000
TSCA Yes