Learn More
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Supplier: Thermo Scientific Chemicals 168600010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium methoxide | |
| 124-41-4,67-56-1 | |
| Colorless to Yellow | |
| 93.0°C | |
| Glass Bottle | |
| 1.3701 | |
| MFCD00012179 | |
| 01,28 | |
| 0.97 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| sodium;methanolate | |
| 10942334 | |
| 54.02 | |
| Liquid |
| 30 wt% Solution in Methanol | |
| 1.0°C to 5.0°C | |
| 0.9700g/mL | |
| 33°C | |
| CH3NaO | |
| CH3ONa | |
| 1 L | |
| 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352 | |
| 15,8775 | |
| Solubility in water: reacts. Other solubilities: soluble in methanol | |
| C[O-].[Na+] | |
| 54.02 | |
| 68 mPa.s (20°C) | |
| Pure |
Chemical Identifiers
| 124-41-4 | |
| 54.02 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| 10942334 | |
| C[O-].[Na+] |
| CH3NaO | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| sodium;methanolate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Flammable liquid and vapor.
May be corrosive to metals.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 204-699-5
RUO – Research Use Only