missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methoxide (ca. 5mol/L in Methanol), TCI America™

Catalog No. S0486100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0486100ML 100 mL
S0486500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0486100ML Supplier TCI America Supplier No. S0486100ML

Chemical Identifiers

CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
UN Number 1289
Formula Weight 54.02
Chemical Name or Material Sodium Methoxide (ca. 5mol/L in Methanol)
EINECSNumber (2)-0203
RTECSNumber PC3570000
TSCA Yes