Learn More
Sodium methoxide, 98%
CAS: 124-41-4 | CH3NaO | 54.024 g/mol
Supplier: Thermo Scientific Chemicals L0567336
Description
Sodium methoxide is used as a base and a reagent in organic chemistry. It is useful in the synthesis of pharmaceuticals and agrochemicals. It acts as a nucleophile and is used in the production of methyl ethers. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight. It is involved in the production of biodiesel from vegetable oils and animal fats.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium methoxide is used as a base and a reagent in organic chemistry. It is useful in the synthesis of pharmaceuticals and agrochemicals. It acts as a nucleophile and is used in the production of methyl ethers. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight. It is involved in the production of biodiesel from vegetable oils and animal fats.
Solubility
Miscible with ethanol, methanol, fats and esters.Immiscible with hydrocarbons and organic solvents.
Notes
Air, moisture and heat sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents, alkali metals, chlorinated solvents, water and reducing agents.
Specifications
| Sodium methoxide | |
| >300°C | |
| CH3ONa | |
| 500 g | |
| 3592982 | |
| 14,8643 | |
| Miscible with ethanol,methanol,fats and esters.Immiscible with hydrocarbons and organic solvents. | |
| C[O-].[Na+] | |
| 54.024 | |
| 54.02 |
| 124-41-4 | |
| CH3NaO | |
| MFCD00012179 | |
| UN1431 | |
| Air and moisture sensitive | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| sodium;methanolate | |
| 10942334 | |
| 98% |
Chemical Identifiers
| 124-41-4 | |
| 54.024 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| 10942334 | |
| C[O-].[Na+] |
| CH3NaO | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| sodium;methanolate |
Safety and Handling
GHS H Statement
H228-H251-H314-H318
Flammable solid.
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P234-P235+P410-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
H228-H290-H302-H314-H335-H500-X
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: SODIUM METHYLATE
EINECSNumber : 204-699-5
RTECSNumber : PC3570000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only