missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, 98%

Catalog No. AAL056730P
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL056730P 2000 g
AAL0567322 100 g
AAL0567336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL056730P Supplier Thermo Scientific Chemicals Supplier No. L056730P
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-41-4 | CH3NaO | 54.024 g/mol

Sodium methoxide is used as a base and a reagent in organic chemistry. It is useful in the synthesis of pharmaceuticals and agrochemicals. It acts as a nucleophile and is used in the production of methyl ethers. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight. It is involved in the production of biodiesel from vegetable oils and animal fats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium methoxide is used as a base and a reagent in organic chemistry. It is useful in the synthesis of pharmaceuticals and agrochemicals. It acts as a nucleophile and is used in the production of methyl ethers. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight. It is involved in the production of biodiesel from vegetable oils and animal fats.

Solubility
Miscible with ethanol, methanol, fats and esters.Immiscible with hydrocarbons and organic solvents.

Notes
Air, moisture and heat sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents, alkali metals, chlorinated solvents, water and reducing agents.

Chemical Identifiers

CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point >300°C
Linear Formula CH3ONa
Quantity 2000 g
UN Number UN1431
Beilstein 3592982
Sensitivity Air and moisture sensitive
Merck Index 14,8643
Solubility Information Miscible with ethanol,methanol,fats and esters.Immiscible with hydrocarbons and organic solvents.
Formula Weight 54.02
Percent Purity 98%
Chemical Name or Material Sodium methoxide
Show More Show Less
Hazard Category H228-H290-H302-H314-H335-H500-X
Hazard Statement GHS H Statement
H228-H251-H314-H318
Flammable solid.
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P234-P235+P410-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: SODIUM METHYLATE
EINECSNumber 204-699-5
RTECSNumber PC3570000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only