missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, 0.5M solution in methanol, AcroSeal™

Catalog No. p-4089436
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427221000 100 mL AcroSeal™ Glass Bottle
AC427228000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427221000 Supplier Thermo Scientific Chemicals Supplier No. 427221000
Only null left

CAS: 124-41-4 | CH3NaO | 54.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-41-4 , 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Boiling Point 65°C
Melting Point -98°C
Color Colorless
Physical Form Liquid
Quantity 100 mL
Linear Formula CH3ONa
Flash Point 11°C
Merck Index 15, 8775
Solubility Information Solubility in water: reacts
Formula Weight 54.02
Concentration or Composition (by Analyte or Components) 0.45 to 0.60M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.8
Density 0.8000g/mL
Chemical Name or Material Sodium methoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
EINECSNumber 204-699-5

RUO – Research Use Only