missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7136014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S098725G 25 g
S0987100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S098725G Supplier TCI America Supplier No. S098725G
Only null left

Chemical Identifiers

CAS 5536-61-8
Molecular Formula C4H5NaO2
Molecular Weight (g/mol) 108.07
MDL Number MFCD00045886
InChI Key SONHXMAHPHADTF-UHFFFAOYSA-M
Synonym sodium methacrylate, sodium polymethacrylate, acrynax, kometa, comet, komet, osakryl s, darvan 7, daxad 30, pma-na
PubChem CID 3255932
IUPAC Name sodium 2-methylprop-2-enoate
SMILES [Na+].CC(=C)C([O-])=O

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 108.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1027
TSCA Yes