accessibility menu, dialog, popup

UserName

CAS RN 5536-61-8

NaH3CCH2OO

CAS RN 5536-61-8

IUPAC Name: sodium 2-methylprop-2-enoate
Synonyms: sodium polymethacrylate
Molecular Weight (g/mol): 108.07
Molecular Formula: C4H5NaO2
InChi Key: SONHXMAHPHADTF-UHFFFAOYSA-M
SMILES: [Na+].CC(=C)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results