missing translation for 'onlineSavingsMsg'
Learn More

Sodium meta-Periodate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-34072
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S39850 50 g Poly Bottle
S398100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S39850 Supplier Thermo Fisher Scientific Supplier No. S39850
Only null left

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 300°C
Color White
pH 3.4 to 4.5
Physical Form Solid
Quantity 50 g
Assay Percent Range 99.8 to 100.3 %
Formula Weight 213.89
Percent Purity 99.8 to 100.3%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium meta-Periodate
Show More Show Less
DOTInformation DOT Class 5.1, : Oxidizer