missing translation for 'onlineSavingsMsg'
Learn More

Sodium Mesitylenesulfonate 98.0+%, TCI America™

Catalog No. M079625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M079625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M079625G Supplier TCI America Supplier No. M079625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6148-75-0
Molecular Formula C9H11NaO3S
Molecular Weight (g/mol) 222.23
MDL Number MFCD00067712
InChI Key AOJUNZYQOYSGHT-UHFFFAOYSA-M
Synonym sodium mesitylenesulfonate, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt, sodium 2,4,6-trimethylbenzenesulfonate, sodium 2,4,6-trimethylbenzenesulphonate, mesitylenesulfonic acid sodium salt hemihydrate, mesitylenesulfonic acid sodium salt, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1, 2,4,6-trimethylbenzenesulfonic acid sodium salt, mesitylene sodium sulfonate, acmc-1bbae
PubChem CID 23661859
IUPAC Name sodium 2,4,6-trimethylbenzene-1-sulfonate
SMILES [Na+].CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 222.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Mesitylenesulfonate