missing translation for 'onlineSavingsMsg'
Learn More

Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical™

Catalog No. p-192571
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S529500 500 g Glass Bottle
S5293 3 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S529500 Supplier Thermo Fisher Scientific Supplier No. S529 500
Only null left

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium Dodecyl Sulfate, SDS, Dodecylsulfuric Acid Sodium Salt, Duponol™ C Dry
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Solid
Melting Point 206°C
pH 8.5 to 10
Quantity 500 g
Flash Point 100°C
Identification Pass Test (FCC), Pass Test (NF)
Packaging Glass Bottle
Linear Formula CH3(CH2)10CH2OSO3Na
UN Number 1325
Grade FCC/NF
Chemical Name or Material Sodium Lauryl Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 4.1, : Flammable Solid