missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lauryl Sulfate, NF, EP, BP, JP, Spectrum™ Chemical
SO180, 151-21-3, C12H25O4SNa
$288.53 - $2694.00
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| IUPAC Name | sodium dodecyl sulfate |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606616
|
Spectrum Chemical Mfg Cor
SO180125GM |
125 g | Poly Bottle |
Each for $288.53
|
|
||||
|
18606617
|
Spectrum Chemical Mfg Cor
SO180500GM |
500 g | Poly Bottle |
Each for $665.30
|
|
||||
|
18606618
|
Spectrum Chemical Mfg Cor
SO1801KG |
1 kg | Poly Bottle |
Each for $1,213.53
|
|
||||
|
18606619
|
Spectrum Chemical Mfg Cor
SO18025KG |
2.5 kg | Poly Bottle |
Each for $2,694.00
|
|
||||
Description
Spectrum™ Chemical Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate |
Specifications
| 151-21-3 | |
| Slightly Alkaline | |
| Poly Bottle | |
| 0.05 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| ≥ 85% | |
| Sodium Lauryl Sulfate |
| 1 | |
| 125 g | |
| C12H25NaO4S | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| Multi-Compendial |