Learn More
Sodium Lauryl Sulfate, FCC, 59%, Spectrum™ Chemical
S1332, 151-21-3, C12H25O4SNa
Supplier: Spectrum Chemical Mfg Cor S133212KG
| Quantity | 12 kg |
|---|---|
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical Sodium Lauryl Sulfate, FCC is a surfactant and emulsifier commonly used in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate |
Specifications
| Sodium Lauryl Sulfate | |
| 1 | |
| Poly Bottle | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| FCC |
| 151-21-3 | |
| 12 kg | |
| C12H25NaO4S | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 59% |