missing translation for 'onlineSavingsMsg'
Learn More

Sodium Lauryl Sulfate, FCC, 59%, Spectrum™ Chemical

S1332, 151-21-3, C12H25O4SNa

Supplier:  Spectrum Chemical Mfg Cor S13322.5KG

Quantity 2.5 kg
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18606613


Only null left

Description

Description

Spectrum™ Chemical Sodium Lauryl Sulfate, FCC is a surfactant and emulsifier commonly used in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
C12H25NaO4S
MFCD00036175
sodium dodecyl sulfate

Specifications

Sodium Lauryl Sulfate
1
Poly Bottle
MFCD00036175
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
FCC
151-21-3
2.5 kg
C12H25NaO4S
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
59%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions