missing translation for 'onlineSavingsMsg'
Learn More

Sodium Lauryl Sulfate, FCC, 59%, Spectrum™ Chemical

Catalog No. 18606611
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606611 125 g Poly Bottle
18606612 500 g Poly Bottle
18606613 2.5 kg Poly Bottle
18606614 12 kg Poly Bottle
18606615 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606611 Supplier Spectrum Chemical Mfg Cor Supplier No. S1332125GM

S1332, 151-21-3, C12H25O4SNa

Spectrum™ Chemical Sodium Lauryl Sulfate, FCC is a surfactant and emulsifier commonly used in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Percent Purity 59%
Grade FCC
Chemical Name or Material Sodium Lauryl Sulfate