missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-lactate, 98+%

CAS: 867-56-1 | C3H5NaO3

$127.63 - $423.67

Chemical Identifiers

CAS 867-56-1
Molecular Formula C3H5NaO3
MDL Number MFCD00066576
Synonym sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1450006
View Documents
Thermo Scientific Chemicals
L1450006
5 g
Each for $127.63
Only null left
 
AAL1450014
View Documents
Thermo Scientific Chemicals
L1450014
25 g
Each for $423.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

867-56-1
C3H5NaO3
MFCD00066576
sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt

Specifications

∼160°C
5 g
MFCD00066576
sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt
112.06
Hygroscopic
867-56-1
C3H5NaO3
4567087
Soluble in water.
≥98%
Sodium L-lactate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-762-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only