missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-lactate, 98+%

CAS: 867-56-1 | C3H5NaO3

Supplier:  Thermo Scientific Chemicals L1450006

 View more versions of this product

Catalog No. AAL1450006

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium L-lactate is used as a food additive, preservative, acidity regulator and bulking agent. It finds application in shampoo and liquid soaps and other related products. It acts as an effective humectant and moisturizer. It is used in the treatment of arrhythmias.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium L-lactate
867-56-1
C3H5NaO3
4567087
Soluble in water.
≥98%
∼160°C
5 g
MFCD00066576
sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt
112.06
Hygroscopic

Chemical Identifiers

867-56-1
C3H5NaO3
MFCD00066576
sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-762-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only