missing translation for 'onlineSavingsMsg'
Learn More

Sodium KAP Analysis Standard with ISA, 0.1M, Reagents

Catalog No. 01333941
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333941 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333941 Supplier Labchem Inc Supplier No. CS0139001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium KAP Analysis Standard with ISA, 0.1M, Reagent, use for Qualitative analysis of cations and anions. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium KAP Analysis Standard with ISA, 0.1M
Packaging Poly Bottle
CAS 7732-18-5, 7647-14-5, 1336-21-6
Specific Gravity 1.0054
Physical Form Liquid
Technique Qualitative Analysis of Cations and Anions
Color Colorless
Boiling Point 100°C
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less