missing translation for 'onlineSavingsMsg'
Learn More

Sodium Isonicotinate 99.0+%, TCI America™

Catalog No. p-7134888
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P099125G 25 g
P0991250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P099125G Supplier TCI America Supplier No. P099125G
Only null left

Chemical Identifiers

CAS 16887-79-9
Molecular Formula C6H4NNaO2
Molecular Weight (g/mol) 145.093
MDL Number MFCD00064868
InChI Key PVXCGWNPDFFPOV-UHFFFAOYSA-M
Synonym Isonicotinic Acid Sodium Salt, Pyridine-4-carboxylic Acid Sodium Salt, Sodium Pyridine-4-carboxylate
PubChem CID 23662338
IUPAC Name sodium;pyridine-4-carboxylate
SMILES C1=CN=CC=C1C(=O)[O-].[Na+]

Specifications

Color White
pH 7.0 to 10.0 (50g/L H2O 25°C)
Quantity 25 g
Formula Weight 145.09
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Isonicotinate
RTECSNumber NS2100000
TSCA Yes