accessibility menu, dialog, popup

UserName

NaOON

CAS RN 16887-79-9

IUPAC Name: sodium pyridine-4-carboxylate
Synonyms: sodium pyridine-4-carboxylate
Molecular Weight (g/mol): 145.09
Molecular Formula: C6H4NNaO2
InChi Key: PVXCGWNPDFFPOV-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results