missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Isonicotinate 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| pH | 7.0 to 10.0 (50g/L H2O 25°C) |
| Quantity | 25 g |
| Formula Weight | 145.09 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium Isonicotinate |
Safety and Handling
| RTECSNumber | NS2100000 |
| TSCA | Yes |