missing translation for 'onlineSavingsMsg'
Learn More

Sodium Isoascorbate Monohydrate 98.0+%, TCI America™

Catalog No. I023225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I023225G 25 g
I0232500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I023225G Supplier TCI America Supplier No. I023225G

Chemical Identifiers

CAS 6381-77-7
Molecular Formula C6H8NaO6
Molecular Weight (g/mol) 199.114
MDL Number MFCD00067298
InChI Key WTTKTIBMFBIUSE-RKJRWTFHSA-N
Synonym sodium erythorbate
PubChem CID 66952495
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one;sodium
SMILES C(C(C1C(=C(C(=O)O1)O)O)O)O.[Na]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 198.11
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium Isoascorbate Monohydrate
EINECSNumber (9)-0132
RTECSNumber MP8910000
TSCA Yes