missing translation for 'onlineSavingsMsg'
Learn More

Sodium Isethionate 98.0+%, TCI America™

Catalog No. p-7130433
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H024125G 25 g
H0241500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H024125G Supplier TCI America Supplier No. H024125G

Chemical Identifiers

CAS 1562-00-1
Molecular Formula C2H5NaO4S
Molecular Weight (g/mol) 148.108
MDL Number MFCD00007534
InChI Key LADXKQRVAFSPTR-UHFFFAOYSA-M
Synonym sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate
PubChem CID 517063
IUPAC Name sodium;2-hydroxyethanesulfonate
SMILES C(CS(=O)(=O)[O-])O.[Na+]

Specifications

Melting Point 194°C
Color White
Quantity 25 g
Formula Weight 148.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Isethionate
EINECSNumber (2)-1646
TSCA Yes