accessibility menu, dialog, popup

UserName

NaOHSOOO

CAS RN 1562-00-1

IUPAC Name: sodium 2-hydroxyethane-1-sulfonate
Synonyms: sodium isethionate
Molecular Weight (g/mol): 148.11
Molecular Formula: C2H5NaO4S
InChi Key: LADXKQRVAFSPTR-UHFFFAOYSA-M
SMILES: [Na+].OCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)