missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodate, 99% min

CAS: 7681-55-2 | INaO3 | 197.891 g/mol

$106.79 - $374.25

Chemical Identifiers

CAS 7681-55-2
Molecular Formula INaO3
Molecular Weight (g/mol) 197.891
MDL Number MFCD00003533
InChI Key WTCBONOLBHEDIL-UHFFFAOYSA-M
Synonym sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
PubChem CID 23675764
ChEBI CHEBI:81708
IUPAC Name sodium;iodate
SMILES [O-]I(=O)=O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4013522
View Documents
Thermo Scientific Chemicals
04013522
100 g
Each for $106.79
Only null left
 
AA4013536
View Documents
Thermo Scientific Chemicals
04013536
500 g
Each for $374.25
Only null left
 
Description

Description

Sodium iodate is used in animal feed, food industry. It is utilized in the manufacture of disinfectants, antiseptics and deodorants. It acts as a significant part in the oxidation of aromatic, aliphatic, and alicyclic thiols. Potassium tetrahydroborate and sodium iodate in acidic aqueous solution react with cadmium to generate chemical vapor, which is useful for atomic fluorescence spectrometry application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7681-55-2
197.891
WTCBONOLBHEDIL-UHFFFAOYSA-M
23675764
sodium;iodate
INaO3
MFCD00003533
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
CHEBI:81708
[O-]I(=O)=O.[Na+]

Specifications

(decomposition)
White
100 g
MFCD00003533
14,8630
Soluble in water and acetone.
[O-]I(=O)=O.[Na+]
197.891
CHEBI:81708
≤99%
4.28 g/mL
7681-55-2
Crystalline Powder
INaO3
UN1479
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium;iodate
23675764
197.89
Light and moisture sensitive
Sodium iodate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H302-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.

P210-P220-P221-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c

H272-H302-H317

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 231-672-5

RTECSNumber : NN1400000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only