missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodate, 99% min

Catalog No. AA4013536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4013536 500 g
AA4013522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4013536 Supplier Thermo Scientific Chemicals Supplier No. 04013536

CAS: 7681-55-2 | INaO3 | 197.891 g/mol

Sodium iodate is used in animal feed, food industry. It is utilized in the manufacture of disinfectants, antiseptics and deodorants. It acts as a significant part in the oxidation of aromatic, aliphatic, and alicyclic thiols. Potassium tetrahydroborate and sodium iodate in acidic aqueous solution react with cadmium to generate chemical vapor, which is useful for atomic fluorescence spectrometry application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-55-2
Molecular Formula INaO3
Molecular Weight (g/mol) 197.891
MDL Number MFCD00003533
InChI Key WTCBONOLBHEDIL-UHFFFAOYSA-M
Synonym sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
PubChem CID 23675764
ChEBI CHEBI:81708
IUPAC Name sodium;iodate
SMILES [O-]I(=O)=O.[Na+]

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 500 g
UN Number UN1479
Merck Index 14,8630
Solubility Information Soluble in water and acetone.
Formula Weight 197.89
Percent Purity ≤99%
Sensitivity Light and moisture sensitive
Density 4.28 g/mL
Chemical Name or Material Sodium iodate
Show More Show Less
Hazard Category H272-H302-H317
Hazard Statement GHS H Statement
H272-H334-H302-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 231-672-5
RTECSNumber NN1400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only