missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodate, 99+%, for analysis

CAS: 7681-55-2 | INaO3 | 197.89 g/mol

$111.31 - $111.31

Chemical Identifiers

CAS 7681-55-2
Molecular Formula INaO3
Molecular Weight (g/mol) 197.89
MDL Number MFCD00003533
InChI Key WTCBONOLBHEDIL-UHFFFAOYSA-M
Synonym sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
PubChem CID 23675764
ChEBI CHEBI:81708
IUPAC Name sodium;iodate
SMILES [O-]I(=O)=O.[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC201761000
SDS View Documents
Thermo Scientific Chemicals
201761000
100 g Plastic bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7681-55-2
197.89
WTCBONOLBHEDIL-UHFFFAOYSA-M
23675764
sodium;iodate
INaO3
MFCD00003533
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
CHEBI:81708
[O-]I(=O)=O.[Na+]

Specifications

560.0°C
White
100 g
INaO3
MFCD00003533
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium;iodate
23675764
197.89
Analytical
Plastic bottle
7681-55-2
Fine Crystalline Powder
99% min. (Iodometry)
NaIO3
15, 8764
Solubility in water: soluble. Other solubilities: soluble in acetone, insoluble in alcohol
[O-]I(=O)=O.[Na+]
197.89
CHEBI:81708
≥99%
0.1% max. (130°C)
Sodium iodate, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 231-672-5

RTECSNumber : NN1400000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only