missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ibandronate 90.0+%, TCI America™

Catalog No. p-7135975
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08771G 1 g
S08775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08771G Supplier TCI America Supplier No. S08771G
Only null left

Chemical Identifiers

CAS 138844-81-2
Molecular Formula C9H22NNaO7P2
Molecular Weight (g/mol) 341.21
MDL Number MFCD07197214
InChI Key LXLBEOAZMZAZND-UHFFFAOYNA-M
Synonym ibandronate sodium, bondronat, boniva, ibandronate sodium salt, sodium ibandronate, 1-hydroxy-3-methylpentylamino propylidene bisphosphonic acid sodium, phosphonic acid, 1-hydroxy-3-methylpentylamino propylidene bis-, monosodium salt, sodiumibandronate, ibandronic acid sodium salt
PubChem CID 23670359
IUPAC Name sodium hydrogen {1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl}phosphonate
SMILES [Na+].CCCCCN(C)CCC(O)(P(O)(O)=O)P(O)([O-])=O

Specifications

Color White
Quantity 1 g
Formula Weight 341.21
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ibandronate
RTECSNumber SZ8563300
TSCA No
Recommended Storage Refrigerator