accessibility menu, dialog, popup

UserName

H3CNCH3OHPOHOOHPOHOONa

CAS RN 138844-81-2

IUPAC Name: sodium hydrogen {1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl}phosphonate
Synonyms: sodium hydrogen 1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropylphosphonate
Molecular Weight (g/mol): 341.21
Molecular Formula: C9H22NNaO7P2
InChi Key: LXLBEOAZMZAZND-UHFFFAOYNA-M
SMILES: [Na+].CCCCCN(C)CCC(O)(P(O)(O)=O)P(O)([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results