missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, Reagent grade, Available chlorine 4.00 to 4.99%, Honeywell
Reagent Grade, available chlorine 4.00-4.99Â %
$1,800.58 - $1,800.58
Chemical Identifiers
| CAS | 7681-52-9 |
|---|---|
| Molecular Formula | ClNaO |
| Molecular Weight (g/mol) | 74.439 |
| MDL Number | MFCD00011120 |
| InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
| Synonym | sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution |
| PubChem CID | 23665760 |
| ChEBI | CHEBI:32146 |
| IUPAC Name | sodium;hypochlorite |
| SMILES | [O-]Cl.[Na+] |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 7681-52-9 | |
| 74.439 | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| 23665760 | |
| sodium;hypochlorite |
| ClNaO | |
| MFCD00011120 | |
| sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
| CHEBI:32146 | |
| [O-]Cl.[Na+] |
Specifications
| 7681-52-9 | |
| ClNaO | |
| MFCD00011120 | |
| sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
| [O-]Cl.[Na+] | |
| 74.439 | |
| CHEBI:32146 | |
| Reagent | |
| 17.5mmHg (20°C) | |
| Sodium Hypochlorite Solution |
| 5 L | |
| NaClO | |
| 3082 | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| sodium;hypochlorite | |
| 23665760 | |
| 74.44g/mol | |
| Available chlorine 4.00 to 4.99% | |
| 1.097g/mL at 25°C |
Safety and Handling
P273-P280-P305 + P351 + P338
H315-H318-H400
EINECSNumber : 231-668-3