missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, Reagent grade, Available chlorine 4.00 to 4.99%, Solstice
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 5 L |
| Linear Formula | NaClO |
| UN Number | 3082 |
| Formula Weight | 74.44g/mol |
| Grade | Reagent |
| Concentration or Composition (by Analyte or Components) | Available chlorine 4.00 to 4.99% |
| Vapor Pressure | 17.5mmHg (20°C) |
| Density | 1.097g/mL at 25°C |
| Chemical Name or Material | Sodium Hypochlorite Solution |
Safety and Handling
| Hazard Statement | P273-P280-P305 + P351 + P338 |
| Precautionary Statement | H315-H318-H400 |
| EINECSNumber | 231-668-3 |