missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, 5% available Chlorine, Ricca Chemical
$63.53 - $2,241.06
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Weight (g/mol) | 74.44 g/mol |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7495-516
|
Ricca Chemical Company
7495516 |
500 mL | Amber Poly Bottle |
N/A
|
|
||||
|
7495-532
|
Ricca Chemical Company
7495532 |
1 L | Amber Poly Bottle |
N/A
|
|
||||
|
7495-51
|
Ricca Chemical Company
749551 |
4 L | Black Poly Bottle |
N/A
|
|
||||
|
7495-555
|
Ricca Chemical Company
7495555 |
55 gal. | Poly Drum |
N/A
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 74.44 g/mol | |
| dihydrogen oxide, dihydrogen monoxide |
Specifications
| Colorless to Green-Yellow | |
| 92.0,5.88 | |
| Liquid | |
| dihydrogen oxide, dihydrogen monoxide | |
| 5% W/W | |
| 5% available Chlorine | |
| Sodium Hypochlorite Solution |
| 7732-18-5,7681-52-9,7732-18-5 | |
| 96.0,6.12 | |
| 500 mL | |
| 74.44 g/mol | |
| Laboratory | |
| Amber Poly Bottle |
Safety and Handling
ShelfLife : 6 months