missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, 2.5% (w/w) NaOCl, Ricca Chemical

Catalog No. 7495116
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7495116 500 mL Natural Poly Bottle
7495132 1 L Natural Poly Bottle
749511 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7495116 Supplier Ricca Chemical Company Supplier No. 7495.116

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Colorless to Green-Yellow
CAS Min % 97.0, 2.15
CAS Max % 99.0, 2.23
Physical Form Liquid
Quantity 500 mL
Percent Purity 2.5% W/W
Grade Laboratory
Concentration or Composition (by Analyte or Components) 2.5% (w/w) NaOCl
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hypochlorite Solution
ShelfLife 6 months