missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Solution, 12% Available Chlorine, Reagent, Spectrum™ Chemical

S1316, 7681-52-9

$1112.37 - $4389.60

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606592
View Documents
Spectrum Chemical Mfg Cor
S1316500MLPLCS6
6 x 500 mL Poly Bottle
Case of 6 Each for $1,112.37
Only null left
 
18606594
View Documents
Spectrum Chemical Mfg Cor
S13164LTCS4
4 x 4 L Poly Bottle
Case of 4 Each for $3,226.57
Only null left
 
18606595
View Documents
Spectrum Chemical Mfg Cor
S1316200LTBL
200 L Drum
Each for $4,389.60
Only null left
 
Description

Description

Spectrum™ Chemical Sodium Hypochlorite, Solution, 12 Percent Available Chlorine, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. It is most commonly used as a bleaching agent to remove stains from laundry in the household. Sodium hypochlorite is also used for disinfection and cleaning.

Specifications

Chemical Identifiers

7681-52-9
74.44
sodium hypochlorite
ClNaO
SUKJFIGYRHOWBL-UHFFFAOYSA-N
[Na+].[O-]Cl

Specifications

7681-52-9,7732-18-5
6 x 500 mL
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium hypochlorite
Poly Bottle
0.13,0.87
ClNaO
[Na+].[O-]Cl
74.44
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'