missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite, Solution, 12% Available Chlorine, Reagent, Spectrum™ Chemical
S1316, 7681-52-9
$250.30 - $1,012.97
Chemical Identifiers
| CAS | 7681-52-9 |
|---|---|
| Molecular Formula | ClNaO |
| Molecular Weight (g/mol) | 74.44 |
| InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
| IUPAC Name | sodium hypochlorite |
| SMILES | [Na+].[O-]Cl |
Description
Spectrum™ Chemical Sodium Hypochlorite, Solution, 12 Percent Available Chlorine, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. It is most commonly used as a bleaching agent to remove stains from laundry in the household. Sodium hypochlorite is also used for disinfection and cleaning.Chemical Identifiers
| 7681-52-9 | |
| 74.44 | |
| sodium hypochlorite |
| ClNaO | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| [Na+].[O-]Cl |
Specifications
| 7681-52-9,7732-18-5 | |
| 500 mL | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| sodium hypochlorite | |
| Poly Bottle |
| 0.13,0.87 | |
| ClNaO | |
| [Na+].[O-]Cl | |
| 74.44 |