missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Solution, 12% Available Chlorine, Reagent, Spectrum™ Chemical

Catalog No. 18606591
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606591 500 mL Poly Bottle
18606593 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606591 Supplier Spectrum Chemical Mfg Cor Supplier No. S1316500MLPL

S1316, 7681-52-9

Spectrum™ Chemical Sodium Hypochlorite, Solution, 12 Percent Available Chlorine, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. It is most commonly used as a bleaching agent to remove stains from laundry in the household. Sodium hypochlorite is also used for disinfection and cleaning.

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl

Specifications

CAS Max % 0.13, 0.87
Quantity 500 mL
Packaging Poly Bottle