missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, Certified, 2.0% to 3.0% (w/v), LabChem™

Catalog No. LC246404
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC246404 4 L
LC246401 500 mL
LC246402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC246404 Supplier LabChem Supplier No. LC246404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Yellow
CAS Max % 97.7, 2.3
Physical Form Liquid
Quantity 4 L
Linear Formula NaOCl
Solubility Information Soluble in water
Formula Weight 74.44
Percent Purity 2.0 to 3.0% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Chlorine; Phosgene
Chemical Name or Material Sodium Hypochlorite
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-668-3
Recommended Storage Room Temperature