missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, 5% Available Chlorine, Solution, ACS, Spectrum™ Chemical

Catalog No. 18613676
Change view
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613676 500 mL Poly Bottle
18613677 4 L Poly Bottle
18613678 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613676 Supplier Spectrum Chemical Mfg Cor Supplier No. S1669500ML
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl

Specifications

CAS Max % 1
Physical Form Solution
Quantity 500 mL
Percent Purity 5%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Hypochlorite, 5 Percent Available Chlorine, Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.